missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, sodium form

Product Code. 11452458
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
Product Code. Quantity unitSize
11452458 500 g 500g
11462458 2500 g 2500g
2 options
This item is not returnable. View return policy

Product Code. 11452458

Brand: Thermo Scientific Alfa Aesar A15795.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Forms a colloidal suspension in water | CAS: 1302-78-9

Bentonite, sodium form is used as a suspending agent, a gel binder, a filler in ceramics and a catalyst support.Sodium bentonite is widely utilized as a natural sealant in recreational ponds, dairy and sewage lagoons and city landfills. Due to its colloidal property, it is employed in drilling mud for oil and gas wells. Due to its swelling property, it provides a self-sealing low permeability barrier. It combines with polymers and enhances the surface modifications, which are used for rheological or sealing performance in geo-environmental applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bentonite, sodium form is used as a suspending agent, a gel binder, a filler in ceramics and a catalyst support.Sodium bentonite is widely utilized as a natural sealant in recreational ponds, dairy and sewage lagoons and city landfills. Due to its colloidal property, it is employed in drilling mud for oil and gas wells. Due to its swelling property, it provides a self-sealing low permeability barrier. It combines with polymers and enhances the surface modifications, which are used for rheological or sealing performance in geo-environmental applications.

Solubility
Insoluble in water and forms a colloidal solution.

Notes
Forms a colloidal suspension in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-78-9
MDL Number MFCD00130611

Specifications

Quantity 500 g
Solubility Information Insoluble in water and forms a colloidal solution.
Chemical Name or Material Bentonite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.