missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benz[a]anthracene-d12, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Benzo[a]anthracene D12 |
| CAS | 1718-53-2 |
| Molecular Formula | C18 2H12 |
| Quantity | 100 mg |
| Synonym | 1,2-Benzanthracene-d12, Perdeuteriobenz[a]anthracene |
| InChI Formula | InChI=1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D |
| SMILES | [2H]c1c([2H])c([2H])c2c([2H])c3c(c([2H])c([2H])c4c([2H])c([2H])c([2H])c([2H])c34)c([2H])c2c1[2H] |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriobenzo[a]anthracene |
| Molecular Weight (g/mol) | 240.36 |
| Formula Weight | 240.1692 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction