missing translation for 'onlineSavingsMsg'
Learn More

Benz[a]anthracene, TRC

Product Code. 30342404
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342404 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342404 Supplier Toronto Research Chemicals Supplier No. TRCB1835601G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benz[a]anthracene
CAS 56-55-3
Purity Grade Notes HPLC
Molecular Formula C18 H12
Quantity 1 g
Synonym 1,2-Benz[a]anthracene, 1,2-Benzanthracene, 1,2-Benzanthrene, 1,2-Benzoanthracene, 2,3-Benzophenanthrene, Anthracenes, benz(a)anthracenes, Benz(a)anthracenes, Benzanthracene, Benzanthrene, Benzo[a]anthracene, Benzo[b]phenanthrene, Benzoanthracene, NSC 30970, Tetraphene, Benzo(a)anthracene
InChI Formula InChI=1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H
SMILES c1ccc2cc3c(ccc4ccccc34)cc2c1
IUPAC Name benzo[a]anthracene
Molecular Weight (g/mol) 228.29
Formula Weight 228.0939
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.