missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde, 99.5+%, pure, redistilled, AcroSeal™

Product Code. 10420554 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10420554 1 L 1 L
10379772 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420554 Supplier Thermo Scientific Acros Supplier No. 378360010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-52-7 | C7H6O | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-52-7
Molecular Weight (g/mol) 106.12
InChI Key HUMNYLRZRPPJDN-UHFFFAOYSA-N
IUPAC Name benzaldehyde
SMILES O=CC1=CC=CC=C1

Specifications

Melting Point -26.0°C
Density 1.0430 g/mL
Boiling Point 179.0°C
Flash Point 64°C
Assay Percent Range 99.5+%
Packaging AcroSeal™ Glass bottle
Molecular Formula C7H6O
Linear Formula C6H5CHO
Refractive Index 1.5440 to 1.5460
Quantity 1 L
Merck Index 15, 1060
Specific Gravity 1.043
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: miscible with alcohol, ether and most other common, organic solvents
Formula Weight 106.12
Grade Pure
Chemical Name or Material Benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • Benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.