missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium Chloride, 99.63%, MP Biomedicals™

Product Code. 11434612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434612 100 g 100g
11444612 250 g 250g
11454612 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434612 Supplier MP Biomedicals Supplier No. 0219015880
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
Synonym Alkyldimethylbenzylammonium chloride

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 0.9963
Percent Purity ≥95%
Chemical Name or Material Benzalkonium Chloride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzalkonium Chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
Hazard Statement
  • H302+H312+H332-Harmful if swallowed, in contact with skin or if inhaled
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.