missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33

Product Code. 10552041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10552041 1 kg Glass bottle 1kg
10503771 100 g Glass bottle 100g
10504664 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10552041 Supplier Thermo Scientific Acros Supplier No. 215410010
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride

Specifications

Melting Point 40.0°C
Physical Form Solid or Thick Gel
Quantity 1 kg
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC16H33
Merck Index 15, 1061
Solubility Information Solubility in water: 4000g/L (20°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene,almost insoluble in ether
Percent Purity 95.0 to 104.0% (anhydrous basis)
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.98
Density 0.9800g/mL
Chemical Name or Material Benzalkonium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzalkonium chloride, alkyl distribution from C8H17 to C16H33
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.