Learn More
Benzamide, 98+%
Description
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.
Solubility
Soluble in water, methanol, ammonia, pyridine, carbon tetrachloride, carbon disulfide and ethanol. Slightly soluble in benzene and ethyl ether.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 125°C to 129°C |
| Density | 1.341 |
| Boiling Point | 288°C |
| Flash Point | 180°C (356°F) |
| Quantity | 100 g |
| Beilstein | 385876 |
| Merck Index | 14,1060 |
| Solubility Information | Soluble in water,methanol,ammonia,pyridine,carbon tetrachloride,carbon disulfide and ethanol. Slightly soluble in benzene and ethyl ether. |
| Formula Weight | 121.14 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
- Benzamide
- Warning
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- H302-Harmful if swallowed.
- H341-Suspected of causing genetic defects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- MIXTURE LIST-Contains : Benzamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.