missing translation for 'onlineSavingsMsg'
Learn More

Benzamide, 99%

Product Code. 10439450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10439450 100 g Plastic bottle 100g
10666552 500 g Plastic bottle 500g
10760785 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10439450 Supplier Thermo Scientific Acros Supplier No. 159331000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 55-21-0 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 55-21-0
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00007968
InChI Key KXDAEFPNCMNJSK-UHFFFAOYSA-N
Synonym benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide
PubChem CID 2331
ChEBI CHEBI:28179
IUPAC Name benzamide
SMILES C1=CC=C(C=C1)C(=O)N

Spezifikation

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125.0°C to 129.0°C
Color White
Boiling Point 228.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CONH2
Quantity 100 g
Beilstein 09, 195
Merck Index 15, 1062
Solubility Information Solubility in water: 1g/74mL (20°C). Other solubilities: soluble in hot water,soluble in hot benzene,1g/6mL alcohol- 1g/3.3mL pyridine,slightly soluble in ether
Formula Weight 121.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Benzamide, 99%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Benzamide
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Gefahrenhinweis
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.