missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine, 95%, ACROS Organics™

Product Code. 10463160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10463160 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10463160 Supplier Thermo Scientific Acros Supplier No. 433050100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 618-39-3
CAS Min % 94.0
CAS Max % 100.0
Melting Point 65°C to 70°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C7H8N2
MDL Number MFCD00042826
Quantity 10 g
Synonym benzamidine, benzimidamide, phenylamidine, unii-kue3zy3j1f, ccris 2952, benzamidine protonated, kue3zy3j1f, chembl20936, 1bra, 1oss
InChI Key PXXJHWLDUBFPOL-UHFFFAOYSA-N
SMILES NC(=N)C1=CC=CC=C1
IUPAC Name benzenecarboximidamide
Molecular Weight (g/mol) 120.16
PubChem CID 2332
ChEBI CHEBI:41033
Formula Weight 120.15
Percent Purity ≥94.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Benzamidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.