missing translation for 'onlineSavingsMsg'
Learn More

Melford Benzamidine HCl

Product Code. 30018066
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018066 100 g 100g
30018067 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018066 Supplier Melford Supplier No. B12000100.0
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes.

Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. Target enzymes include trypsin, hymotrypsin, kallikzein and plasmin.

Specifications

Chemical Name or Material Benzamidine HCl
CAS 1670-14-0
Quantity 100 g
Molecular Formula C7H8N2-HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.