missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride Hydrate, 98%

Product Code. 10000550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10000550 25 g Plastic bottle 25g
10581554 5 g Glass Bottle 5g
10549630 100 g Plastic bottle 100g
10247163 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10000550 Supplier Thermo Scientific Acros Supplier No. 105240250
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206752-36-5 , 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
SMILES Cl.NC(=N)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 86°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (on dry substance) (Argentometry)
Packaging Plastic bottle
Linear Formula HCl·xH2O
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 156.61
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzamidine hydrochloride hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Benzamidine hydrochloride hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.