missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride Hydrate (White Crystalline Powder), Fisher BioReagents

Product Code. 10619363
Change view
Click to view available options
:
Poly Bottle; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10619363 Poly Bottle; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10619363 Supplier Fisher BioReagents Supplier No. BP43525
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Benzamidine Hydrochloride is commonly used as a protease inhibitor.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1670-14-0, 206752-36-5
Melting Point 167°C
Color White
Assay Percent Range ≥99.0 %
Packaging Poly Bottle
Molecular Formula C7H9ClN2
Quantity 25 g
Synonym benzamidine hydrochloride, benzimidamide hydrochloride, benzenecarboximidamide hydrochloride, benzenecarboximidamide, monohydrochloride, benzamidinium chloride, benzamidine hcl, benzamidine, hydrochloride, amidinobenzene hydrochloride, benzamidine, monohydrochloride, amidinobenzene hydrochloride 1:1
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=N)N.Cl
IUPAC Name benzenecarboximidamide;hydrochloride
Molecular Weight (g/mol) 156.613
PubChem CID 80289
Formula Weight 156.61
Percent Purity ≥99.0%
Physical Form Powder or Solid
Chemical Name or Material Benzamidine Hydrochloride Hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Benzamidine Hydrochloride Hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.