missing translation for 'onlineSavingsMsg'
Learn More

Benzamidoxime, 95%

Product Code. 11362009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11362009 1 g 1g
11372009 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11362009

Brand: Thermo Scientific Alfa Aesar H26068.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 613-92-3 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-92-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00031485 MFCD00474011
InChI Key MXOQNVMDKHLYCZ-UHFFFAOYSA-N
Synonym benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
PubChem CID 7259353
ChEBI CHEBI:83354
IUPAC Name N'-hydroxybenzenecarboximidamide
SMILES N\C(=N/O)C1=CC=CC=C1

Specifications

Melting Point 72°C to 76°C
Quantity 1 g
Formula Weight 136.15
Percent Purity 95%
Chemical Name or Material Benzamidoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.