missing translation for 'onlineSavingsMsg'
Learn More

Benzanthrone, TRC

Product Code. 17952415
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17952415 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17952415 Supplier Toronto Research Chemicals Supplier No. TRCB1205905G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82-05-3
Molecular Formula C17H10O
Quantity 5 g
Synonym 7H-Benz[de]anthracen-7-one, 1,9-Benz-10-anthrone, 7-Oxobenz[de]anthracene, Benzanthrenone, Benzoanthrone, NSC 5189, NSC 631641, Naphthanthrone
InChI Formula InChI=1S/C17H10O/c18-17-14-8-2-1-7-12(14)13-9-3-5-11-6-4-10-15(17)16(11)13/h1-10H
SMILES O=C1c2ccccc2c3cccc4cccc1c34
IUPAC Name benzo[a]phenalen-7-one
Molecular Weight (g/mol) 230.26
Formula Weight 230.0732
Chemical Name or Material Benzanthrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.