missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,3-disulfonic acid disodium salt, 94%

Product Code. 11419847
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11419847

Brand: Thermo Scientific Alfa Aesar B22811.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 831-59-4 | C6H4Na2O6S2 | 282.19 g/mol

Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

Solubility
Soluble in water( 663 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-59-4
Molecular Formula C6H4Na2O6S2
Molecular Weight (g/mol) 282.19
MDL Number MFCD00007492
InChI Key XWPWZOJBTOJEGW-UHFFFAOYSA-L
Synonym disodium 1,3-benzenedisulfonate, disodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, disodium salt, benzene-1,3-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid disodium salt, disodium benzene-1,3-disulphonate, disodium m-benzenedisulfonate, sodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, sodium salt 1:2, m-benzenedisulfonic acid, disodium salt
PubChem CID 71577
IUPAC Name disodium;benzene-1,3-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)S([O-])(=O)=O

Specifications

Density 1.124
Quantity 100 g
Solubility Information Soluble in water( 663g/L at 20°C).
Formula Weight 282.2
Percent Purity 94%
Chemical Name or Material Benzene-1,3-disulfonic acid disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.