missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,3-disulfonic acid disodium salt, 94%

Product Code. 11429847
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429847 500 g 500g
11409847 25 g 25g
11419847 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429847 Supplier Thermo Scientific Alfa Aesar Supplier No. B22811.36
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 831-59-4 | C6H4Na2O6S2 | 282.19 g/mol

Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzene-1,3-disulfonic acid disodium salt is used as pharmaceutical intermediate.

Solubility
Soluble in water( 663 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 831-59-4
Molecular Formula C6H4Na2O6S2
Molecular Weight (g/mol) 282.19
MDL Number MFCD00007492
InChI Key XWPWZOJBTOJEGW-UHFFFAOYSA-L
Synonym disodium 1,3-benzenedisulfonate, disodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, disodium salt, benzene-1,3-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid disodium salt, disodium benzene-1,3-disulphonate, disodium m-benzenedisulfonate, sodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, sodium salt 1:2, m-benzenedisulfonic acid, disodium salt
PubChem CID 71577
IUPAC Name disodium;benzene-1,3-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)S([O-])(=O)=O

Specifications

Density 1.124
Quantity 500 g
Solubility Information Soluble in water( 663g/L at 20°C).
Formula Weight 282.2
Percent Purity 94%
Chemical Name or Material Benzene-1,3-disulfonic acid disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.