missing translation for 'onlineSavingsMsg'
Learn More

Benzene-1,4-dithiocarboxamide, 97%

Product Code. 15448337
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448337 1 g 1g
15458337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448337 Supplier Thermo Scientific Alfa Aesar Supplier No. H51857.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13363-51-4 | C8H8N2S2 | 196.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13363-51-4
Molecular Formula C8H8N2S2
Molecular Weight (g/mol) 196.286
MDL Number MFCD00046866
InChI Key USHPIZCRGQUHGN-UHFFFAOYSA-N
Synonym benzene-1,4-dithiocarboxamide, 1,4-benzenedicarbothioamide, benzene-1,4-bis carbothioamide
PubChem CID 2795175
IUPAC Name benzene-1,4-dicarbothioamide
SMILES C1=CC(=CC=C1C(=S)N)C(=S)N

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 196.28
Percent Purity 97%
Chemical Name or Material Benzene-1,4-dithiocarboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.