missing translation for 'onlineSavingsMsg'
Learn More

Benzeneruthenium(II) chloride, dimer, 97%

Product Code. 15447067
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15447067

Brand: Thermo Scientific Alfa Aesar H34138.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 37366-09-9 | C12H12Cl4Ru2 | 500.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 37366-09-9
Melting Point 250°C (decomposition)
Molecular Formula C12H12Cl4Ru2
MDL Number MFCD00064686
Quantity 1 g
Synonym benzeneruthenium ii chloride dimer, dibenzenetetrachlorodiruthenium, bis benzene dichlororuthenium, benzene dichlororuthenium dimer, benzeneruthenium dichloride dimer, bis benzene tetrachlorodiruthenium, rucl2 benzene 2, dichloro benzene rutheniumdimer, benzene ruthenium cento chloride
InChI Key YGXMUPKIEHNBNQ-UHFFFAOYSA-J
SMILES C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl
IUPAC Name benzene;dichlororuthenium
Molecular Weight (g/mol) 500.168
PubChem CID 10962144
Formula Weight 500.18
Percent Purity 97%
Chemical Name or Material Benzeneruthenium(II) chloride, dimer
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.