missing translation for 'onlineSavingsMsg'
Learn More

Benzeneseleninic anhydride, 98%

Product Code. 10455105 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10455105 25 g Glass bottle 25g
10664712 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10455105 Supplier Thermo Scientific Acros Supplier No. 199710250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17697-12-0 | C12H10O3Se2 | 360.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17697-12-0
Molecular Formula C12H10O3Se2
Molecular Weight (g/mol) 360.13
MDL Number MFCD00001991
InChI Key FHPZOWOEILXXBD-UHFFFAOYSA-N
Synonym benzeneseleninic anhydride, benzeneseleninic acid anhydride, benzeneseleninyl benzeneseleninate, bis phenylseleninic anhydride, benzeneseleninic acid, anhydride, bis benzeneseleninic anhydride, 1,3-diphenyldiselenoxane 1,3-dioxide #
PubChem CID 87253
IUPAC Name phenylseleninyl benzeneseleninate
SMILES C1=CC=C(C=C1)[Se](=O)O[Se](=O)C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 170°C to 173°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [C6H5Se(O)]2O
Quantity 25 g
UN Number 3283
Fieser 06,240; 07,139; 08,29; 09,32; 10,22; 11,37; 15,18
Formula Weight 360.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzeneseleninic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzeneseleninic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.