missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfinic acid, sodium salt, 97%

Product Code. 10786551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10786551 100 g 100g
10103160 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10786551 Supplier Thermo Scientific Acros Supplier No. 154051000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-55-2 | C6H5NaO2S | 164.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-55-2
Molecular Formula C6H5NaO2S
Molecular Weight (g/mol) 164.15
MDL Number MFCD00013135
InChI Key CHLCPTJLUJHDBO-UHFFFAOYSA-M
Synonym sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate
PubChem CID 2723840
IUPAC Name sodium;benzenesulfinate
SMILES C1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula C6H5SO2Na
Quantity 100 g
Beilstein 11, 2
Fieser 06,526
Formula Weight 164.15
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzenesulfinic acid, sodium salt, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.