missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzenesulfonic acid isopropyl ester, 95%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
A sulfonate ester that acts as a potential effects in drug substances impurity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.
Solubility
Soluble in chloroform and ethyl acetate.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
A sulfonate ester that acts as a potential effects in drug substances impurity.
Solubility
Soluble in chloroform and ethyl acetate.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Color | Colorless to Yellow |
| Refractive Index | 1.502 |
| Quantity | 1 g |
| Beilstein | 1106314 |
| Synonym | isopropyl benzenesulfonate, isopropylbenzenesulfonate, benzenesulfonic acid, 1-methylethyl ester, benzenesulfonic acid isopropyl ester, isopropyl besylate, benzenesulfonic acid,1-methylethyl ester, benzenesulfonic acid 1-methylethyl ester |
| Solubility Information | Soluble in chloroform and ethyl acetate. |
| Formula Weight | 200.25 |
| Percent Purity | 95% |
| Physical Form | Liquid |
| Chemical Name or Material | Benzenesulfonic acid isopropyl ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction