missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl chloride, 99%

Product Code. 10776191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10776191 1 L Glass bottle 1 L
10294760 250 mL Glass bottle 250mL
12153741 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10776191 Supplier Thermo Scientific Acros Supplier No. 148480010
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 98-09-9 | C6H5ClO2S | 176.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-09-9
Molecular Formula C6H5ClO2S
Molecular Weight (g/mol) 176.62
MDL Number MFCD00007426
InChI Key CSKNSYBAZOQPLR-UHFFFAOYSA-N
Synonym phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride
PubChem CID 7369
IUPAC Name benzenesulfonyl chloride
SMILES C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 14.5°C
Density 1.3840g/mL
Boiling Point 251.0°C to 252.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SO2Cl
Refractive Index 1.5508 to 1.5528
Quantity 1 L
Beilstein 11,34
Fieser 01,46; 05,22; 14,27
Merck Index 15,1073
Specific Gravity 1.384
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in alcohol and ether
Formula Weight 176.62
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzenesulfonyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.