missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl Chloride, TRC

Product Code. 30341067
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341067 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341067 Supplier Toronto Research Chemicals Supplier No. TRCB19000010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzenesulfonyl Chloride
CAS 98-09-9
Molecular Formula C6H5ClO2S
Quantity 10 g
Synonym Benzene sulfochloride, Benzenesulfonic chloride, Benzenesulphonyl chloride, NSC 2864, Phenylsulfonyl chloride
InChI Formula InChI=1S/C6H5ClO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H
SMILES ClS(=O)(=O)c1ccccc1
IUPAC Name benzenesulfonyl chloride
Molecular Weight (g/mol) 176.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.