missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride, Reagecon™
BIOPHARMA

Product Code. 11414789 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414789 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414789 Supplier Reagecon Supplier No. 3000900
  • ONLINE EXCLUSIVE PRICE
    53.45 €
    44.66 € / 1 L
    Save 8.79 €
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121-54-0
Molecular Formula C27H42ClNO2
Quantity 1 L
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Molecular Weight (g/mol) 448.09
PubChem CID 8478
ChEBI CHEBI:31264
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.