missing translation for 'onlineSavingsMsg'
Learn More

Benzil, 98%

Product Code. 11464284
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464284 100 g 100g
11474284 500 g 500g
11484284 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464284 Supplier Thermo Scientific Alfa Aesar Supplier No. A12579.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 134-81-6 | C14H10O2 | 210.23 g/mol

Benzil is used as a pharmaceutical intermediate and uv curing resin photosensitizer. In polymer chemistry, it is used as a photoinitiator. Further, it serves as a potent inhibitor of human carboxylesterases. It is used in the preparation of diketimines by reacting with amines. It is also involved in the famous benil-benzilic acid rearrangement. In addition this, it reacts with 1,3-diphenylacetone to get tetraphenylcyclopentadienone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzil is used as a pharmaceutical intermediate and uv curing resin photosensitizer. In polymer chemistry, it is used as a photoinitiator. Further, it serves as a potent inhibitor of human carboxylesterases. It is used in the preparation of diketimines by reacting with amines. It is also involved in the famous benil-benzilic acid rearrangement. In addition this, it reacts with 1,3-diphenylacetone to get tetraphenylcyclopentadienone.

Solubility
Soluble in ethanol and ethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00003080
InChI Key WURBFLDFSFBTLW-UHFFFAOYSA-N
Synonym benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
PubChem CID 8651
ChEBI CHEBI:51507
IUPAC Name 1,2-diphenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 94°C to 96°C
Density 1.521
Boiling Point 346°C to 348°C
Quantity 100 g
Beilstein 608047
Merck Index 14,1078
Solubility Information Soluble in ethanol and ethyl ether. Insoluble in water.
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material Benzil
compliance-icons
Product Identifier
  • Benzil
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: diphenylethanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.