missing translation for 'onlineSavingsMsg'
Learn More

Benzil, 99+%

Product Code. 10677052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10677052 500 g Plastic bottle 500g
10346070 100 g Plastic bottle 100g
11964841 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10677052 Supplier Thermo Scientific Acros Supplier No. 105415000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 134-81-6 | C14H10O2 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00003080
InChI Key WURBFLDFSFBTLW-UHFFFAOYSA-N
Synonym benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
PubChem CID 8651
ChEBI CHEBI:51507
IUPAC Name 1,2-diphenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 94.0°C to 97.0°C
Color White
Boiling Point 346.0°C to 348.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COCOC6H5
Quantity 500 g
Merck Index 15,108
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in alcohol,ether,chloroform,ethyl,acetate,benzene,toluene and nitrobenzene
Formula Weight 210.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Benzil, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzil
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.