missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole, 98%

Product Code. 10487390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10487390 100 g Plastic bottle 100g
10644882 25 g Plastic bottle 25g
10654882 500 g Plastic bottle 500g
10105303 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10487390 Supplier Thermo Scientific Acros Supplier No. 186421000
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51-17-2 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
IUPAC Name 1H-benzimidazole
SMILES N1C=NC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 174.0°C
Color Beige to Brown
Boiling Point >360.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 23, 131
Merck Index 15, 1083
Solubility Information Solubility in water: sparingly soluble. Other solubilities: more soluble in hot water,sparingly soluble in ether,practically insoluble in benzene and petroleum,soluble in aq. sol. of acids and strong alkalies,freely soluble in alcohol,boiling xylene
Formula Weight 118.14
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzimidazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.