missing translation for 'onlineSavingsMsg'
Learn More

Benzimidazole, 99%

Product Code. 11415103
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415103 50 g 50g
11425103 250 g 250g
11405103 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415103 Supplier Thermo Scientific Alfa Aesar Supplier No. A12763.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-17-2 | C7H6N2 | 118.14 g/mol

Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
SMILES N1C=NC2=CC=CC=C12

Specifications

Melting Point 170°C to 174°C
Boiling Point 360°C
Quantity 50 g
Beilstein 109682
Merck Index 14,1081
Solubility Information Sparingly soluble in water.
IUPAC Name 1H-1,3-benzodiazole
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material Benzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.