missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-boronic acid, 98%

Product Code. 11460088
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460088 25 g 25g
11440088 1 g 1g
11450088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460088 Supplier Thermo Scientific Alfa Aesar Supplier No. B23676.14
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98437-24-2 | C8H7BO3 | 161.95 g/mol

It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

Solubility
Soluble in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98437-24-2
Molecular Formula C8H7BO3
Molecular Weight (g/mol) 161.95
MDL Number MFCD00236019
InChI Key PKRRNTJIHGOMRC-UHFFFAOYSA-N
Synonym benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PubChem CID 2776266
IUPAC Name 1-benzofuran-2-ylboronic acid
SMILES OB(O)C1=CC2=CC=CC=C2O1

Specifications

Quantity 25 g
Beilstein 1637889
Solubility Information Soluble in water.
Formula Weight 161.95
Percent Purity 98%
Chemical Name or Material Benzo[b]furan-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.