missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-7-carboxylic acid, Thermo Scientific™

Product Code. 10488884
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10488884 1 g Amber Glass Bottle 1g
10362095 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10488884 Supplier Thermo Scientific Maybridge Supplier No. CC78201DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 90484-22-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD10000617
InChI Key QMHILIQFOBNARN-UHFFFAOYSA-N
Synonym benzofuran-7-carboxylic acid, benzofuran-7-carboxylicacid, 7-benzofuranylcarboxylic acid, benzo b furan-7-carboxylic acid, 7-benzofurancarboxylic acid, 4ahs, 7-benzofurancarboxylicacid, akh
PubChem CID 13307983
IUPAC Name 1-benzofuran-7-carboxylic acid
SMILES OC(=O)C1=C2OC=CC2=CC=C1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.14
Percent Purity 97%
Chemical Name or Material Benzo[b]furan-7-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.