missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-7-sulfonyl chloride, Thermo Scientific™

Product Code. 10200265
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10200265 1 g Amber Glass Bottle 1g
10670466 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10200265 Supplier Thermo Scientific Maybridge Supplier No. CC78203DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1191030-88-2
Molecular Formula C8H5ClO3S
Molecular Weight (g/mol) 216.635
InChI Key MEOOVUNNRYYGCV-UHFFFAOYSA-N
Synonym benzofuran-7-sulfonyl chloride, 7-benzofuransulfonyl chloride, benzo 2,3-b furan-7-ylchlorosulfone
PubChem CID 49762852
IUPAC Name 1-benzofuran-7-sulfonyl chloride
SMILES C1=CC2=C(C(=C1)S(=O)(=O)Cl)OC=C2

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 216.64
Percent Purity 97%
Chemical Name or Material Benzo[b]furan-7-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.