missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxylic acid, 98%

Product Code. 11464074
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464074 1 g 1g
11474074 5 g 5g
11484074 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464074 Supplier Thermo Scientific Alfa Aesar Supplier No. A12471.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6314-28-9 | C9H6O2S | 178.21 g/mol

Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in ethanol, acetone and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6314-28-9
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00051636
InChI Key DYSJMQABFPKAQM-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxylic acid, thianaphthene-2-carboxylic acid, thionaphthene-2-carboxylic acid, tnca, benzothiophene-2-carboxylic acid, 2-benzo b thiophenecarboxylic acid, unii-hlr49221fq, thionapthene-2-carboxylic acid, thianaphthene-2-carboxylicacid
PubChem CID 95864
IUPAC Name 1-benzothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point 236°C to 242°C
Quantity 1 g
Beilstein 124613
Merck Index 14,9345
Solubility Information Soluble in ethanol,acetone and chloroform. Insoluble in water.
Formula Weight 178.22
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.