missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene-2-methanol, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Benzo[b]thiophene-2-methano is an important intermediates and reagents in organic synthesis.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Benzo[b]thiophene-2-methano is an important intermediates and reagents in organic synthesis.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Especificaciones
| Melting Point | 98°C to 101°C |
| Quantity | 5 g |
| Formula Weight | 164.22 |
| Percent Purity | 97% |
| Chemical Name or Material | Benzo[b]thiophene-2-methanol |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?