missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-acetonitrile, 98+%

Product Code. 11362576
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362576 5 g 5g
11352576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362576 Supplier Thermo Scientific Alfa Aesar Supplier No. L08547.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3216-48-6 | C10H7NS | 173.233 g/mol

Benzo[b]thiophene-3-acetonitrile is used to produce 2-(5-Bromo-benzo[b]thiophen-3-yl)-ethylamine; hydrochloride. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-acetonitrile is used to produce 2-(5-Bromo-benzo[b]thiophen-3-yl)-ethylamine; hydrochloride. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3216-48-6
Molecular Formula C10H7NS
Molecular Weight (g/mol) 173.233
MDL Number MFCD00027389
InChI Key DEELHLMCCWZFFE-UHFFFAOYSA-N
Synonym benzo b thiophene-3-acetonitrile, 2-benzo b thiophen-3-yl acetonitrile, 2-1-benzothiophen-3-yl acetonitrile, thianaphthene-3-acetonirile, 1-benzothien-3-ylacetonitrile, benzo b thiophen-3-yl-acetonitrile, benzo b thiophen-3-yl acetonitrile, 3-cyanomethylbenzothiophene, acmc-1bn87
PubChem CID 76681
IUPAC Name 2-(1-benzothiophen-3-yl)acetonitrile
SMILES C1=CC=C2C(=C1)C(=CS2)CC#N

Specifications

Melting Point 62°C to 66°C
Quantity 5 g
UN Number UN3439
Beilstein 125487
Solubility Information Insoluble in water.
Formula Weight 173.24
Percent Purity ≥98%
Chemical Name or Material Benzo[b]thiophene-3-acetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.