missing translation for 'onlineSavingsMsg'
Learn More

Benzo[d]isoxazol-3-ol, ≥95%, Thermo Scientific™

Product Code. 10191212
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10191212 1 g Amber Glass Bottle 1g
10553052 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10191212 Supplier Thermo Scientific Maybridge Supplier No. RF01278DA
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21725-69-9
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.122
MDL Number MFCD00125030
InChI Key QLDQYRDCPNBPII-UHFFFAOYSA-N
Synonym benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
PubChem CID 210830
IUPAC Name 1,2-benzoxazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NO2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 135.12
Percent Purity 95%
Chemical Name or Material Benzo[d]isoxazol-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.