missing translation for 'onlineSavingsMsg'
Learn More

Benzofuran (1mg/ml in Dichloromethane), TRC

Product Code. 30390767
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390767 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390767 Supplier Toronto Research Chemicals Supplier No. TRCKIT116010X1ML
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 271-89-6
Molecular Formula C8 H6 O
Quantity 10 x 1 mL
Synonym 1-Oxindene, 2,3-Benzofuran, AT 33852, Benzo[b]furan, Benzofurfuran, Coumarone, NSC 1255, R 7204
InChI Formula InChI=1S/C8H6O/c1-2-4-8-7(3-1)5-6-9-8/h1-6H
SMILES o1ccc2ccccc12
IUPAC Name 1-benzofuran
Molecular Weight (g/mol) 118.13
Formula Weight 118.0419
Concentration or Composition (by Analyte or Components) Benzo(b)furan 1 mg/mL
Chemical Name or Material Benzo(b)furan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.