missing translation for 'onlineSavingsMsg'
Learn More

Benzofuran, TRC

Product Code. 30348053
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348053 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348053 Supplier Toronto Research Chemicals Supplier No. TRCB203580100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzo(b)furan
CAS 271-89-6
Purity Grade Notes GC
Molecular Formula C8 H6 O
Quantity 100 g
Synonym 1-Oxindene, 2,3-Benzofuran, AT 33852, Benzo[b]furan, Benzofurfuran, Coumarone, NSC 1255, R 7204
InChI Formula InChI=1S/C8H6O/c1-2-4-8-7(3-1)5-6-9-8/h1-6H
SMILES o1ccc2ccccc12
IUPAC Name 1-benzofuran
Molecular Weight (g/mol) 118.13
Formula Weight 118.0419
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.