missing translation for 'onlineSavingsMsg'
Learn More

Benzofuroxan, 98%

Product Code. 11368255
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368255 10 g 10g
11378255 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368255 Supplier Thermo Scientific Alfa Aesar Supplier No. L10144.09
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 480-96-6 | C6H4N2O2 | 136.11 g/mol

Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-96-6
Molecular Formula C6H4N2O2
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005807
InChI Key OKEAMBAZBICIFP-UHFFFAOYSA-N
Synonym benzofuroxan, benzofuroxane, benzofurazan 1-oxide, benzofurazan oxide, 2,1,3-benzoxadiazole 1-oxide, 2,1,3-benzoxadiazole, 1-oxide, benzofurazan, 1-oxide, 2,1,3-benzoxadiazol-1-ium-1-olate, benzo c 1,2,5-oxadiazol-1-ol, benzofurazan n-oxide
PubChem CID 68075
IUPAC Name 3-oxido-2,1,3-benzoxadiazol-3-ium
SMILES C1=CC2=NO[N+](=C2C=C1)[O-]

Specifications

Melting Point 67°C to 71°C
Quantity 10 g
Beilstein 125112
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 136.11
Percent Purity 98%
Chemical Name or Material Benzofuroxan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.