missing translation for 'onlineSavingsMsg'
Learn More

Benzohydrazide, 98%, BLD Pharm™

Product Code. 31107122
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31107122 100 g
31099528 500 g
31102286 10 g
31103606 25 g
31102008 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31107122 Supplier BLD Pharm Supplier No. BD115613100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 613-94-5
Molecular Formula C7H8N2O
MDL Number MFCD00007596
Quantity 100 g
SMILES O=C(NN)C1=CC=CC=C1
Molecular Weight (g/mol) 136.15
Percent Purity 98%
Chemical Name or Material Benzohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.