missing translation for 'onlineSavingsMsg'
Learn More

Benzohydroxamic acid, 98%

Product Code. 11428763
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428763 10 g 10g
11384589 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428763 Supplier Thermo Scientific Alfa Aesar Supplier No. A10509.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 495-18-1 | C7H7NO2 | 137.138 g/mol

Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

Solubility
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-18-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00002109
InChI Key VDEUYMSGMPQMIK-UHFFFAOYSA-N
Synonym benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
PubChem CID 10313
IUPAC Name N-hydroxybenzamide
SMILES C1=CC=C(C=C1)C(=O)NO

Specifications

Melting Point ∼125°C (decomposition)
Quantity 10 g
Beilstein 1907585
Solubility Information Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material Benzohydroxamic acid
compliance-icons compliance-icons
Product Identifier
  • Benzohydroxamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Benzohydroxamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.