missing translation for 'onlineSavingsMsg'
Learn More

Benzohydroxamic Acid, TRC

Product Code. 30240994
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240994 250 g 250g
30240690 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240994 Supplier Toronto Research Chemicals Supplier No. TRCB203750250G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-18-1
Molecular Formula C7 H7 N O2
Molecular Weight (g/mol) 137.14
Synonym Benzamide, N-hydroxy-, Benzohydroxamic acid (6CI,8CI), N-Hydroxybenzamide, BHAM, Benzenehydroxamic acid, Benzhydroxamic acid, Benzohydroximic acid, Benzoylhydroxylamine, Hydroxylamine, N-benzoyl-, N-Benzohydroxamic acid, N-Benzoylhydroxylamine, NSC 147248, NSC 3136, Phenylhydroxamic acid
IUPAC Name N-hydroxybenzamide
SMILES ONC(=O)c1ccccc1

Specifications

InChI Formula InChI=1S/C7H7NO2/c9-7(8-10)6-4-2-1-3-5-6/h1-5,10H,(H,8,9)
Formula Weight 137.0477
Quantity 250 g
Chemical Name or Material Benzohydroxamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.