missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, ACS Reagent Grade, ≥99.5%, MP Biomedicals™

Product Code. 11480502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480502 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11480502 Supplier MP Biomedicals Supplier No. 0219141280
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 65-85-0
Color White
Density 1.321 g/cm3 (at 20°C)
Quantity 100 g
Physical Form Powder
Assay Percent Range ≥99.5%
Molecular Formula C7H6O2
MDL Number MFCD00002398
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)O
IUPAC Name benzoic acid
Molecular Weight (g/mol) 122.123
PubChem CID 243
ChEBI CHEBI:30746
Formula Weight 122.12
Grade ACS
Chemical Name or Material Benzoic acid, For ACS analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzoic Acid, ACS
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H372-Causes damage to organs.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.