missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, Honeywell Fluka™

Product Code. 16645813 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16645813 1 kg 1kg
16635813 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16645813 Supplier Honeywell Fluka Supplier No. 33047H1KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
PubChem CID 243
ChEBI CHEBI:30746

Specifications

Appearance Complying
Percent Purity 99.9% (min.)
Grade Reagent Pharmacopeia Acids
Quantity 1 kg
Packaging Plastic Bottle
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.