missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, TRC

Product Code. 30011107
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011107 1 g 1g
18081920 50 mg 50mg
18093530 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011107 Supplier Toronto Research Chemicals Supplier No. TRCB2039001G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7 H6 O2
Molecular Weight (g/mol) 122.12
Synonym Benzenecarboxylic acid, Benzeneformic acid, Benzenemethanoic acid, Carboxybenzene, Dracylic acid, E 210, HA 1, HA 1 (acid), MENNO-Florades, NSC 149, Phenylcarboxylic acid, Phenylformic acid, Purox B, Retarder BA, Retardex, Salvo liquid, Solvo powder, Tenn-Plas, VevoVitall, Tiaprofenic Acid Imp. D (EP)
IUPAC Name benzoic acid
SMILES OC(=O)c1ccccc1

Specifications

InChI Formula InChI=1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)
Formula Weight 122.0368
Percent Purity >95%
Quantity 1 g
Chemical Name or Material Benzoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.