missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Anhydride (>90%), TRC

Product Code. 30137821
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137821 100 g 100g
30135197 500 g 500g
30005774 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137821 Supplier Toronto Research Chemicals Supplier No. TRCB203920100G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-97-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
Synonym Benzoic Acid 1,1'-Anhydride, Benzoic Acid Anhydride, Benzoyl Anhydride, Benzoyl Benzoate, Bis(phenylcarbonyl)ether, NSC 37116;
IUPAC Name benzoyl benzoate
SMILES O=C(OC(=O)c1ccccc1)c2ccccc2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C14H10O3/c15-13(11-7-3-1-4-8-11)17-14(16)12-9-5-2-6-10-12/h1-10H
Formula Weight 226.063
Chemical Name or Material Benzoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.