missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-2-carbonyl chloride, 95%

Product Code. 11305425
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305425 250 mg 250mg
11395415 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305425 Supplier Thermo Scientific Alfa Aesar Supplier No. L19631.MD
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 67748-61-2 | C8H4ClNOS | 197.636 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67748-61-2
Molecular Formula C8H4ClNOS
Molecular Weight (g/mol) 197.636
MDL Number MFCD03659697
InChI Key AOIGQLLPWDXVGB-UHFFFAOYSA-N
Synonym benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride
PubChem CID 2776254
IUPAC Name 1,3-benzothiazole-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)N=C(S2)C(=O)Cl

Specifications

Melting Point 113°C to 115°C
Quantity 250 mg
UN Number UN3261
Beilstein 130990
Sensitivity Moisture sensitive
Formula Weight 197.64
Percent Purity 95%
Chemical Name or Material Benzothiazole-2-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.