missing translation for 'onlineSavingsMsg'
Learn More

Benzotriazol-1-yl-oxytripyrrolidinphosphonium Hexafluorophosphate, TRC

Product Code. 30333389
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333389 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333389 Supplier Toronto Research Chemicals Supplier No. TRCB207500100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzotriazol-1-yl-oxytripyrrolidinphosphonium Hexafluorophos
CAS 128625-52-5
Molecular Formula C18 H28 N6 O P . F6 P
Quantity 100 g
Synonym PyBOP
InChI Formula InChI=1S/C18H28N6OP.F6P/c1-2-10-18-17(9-1)19-20-24(18)25-26(21-11-3-4-12-21,22-13-5-6-14-22)23-15-7-8-16-23;1-7(2,3,4,5)6/h1-2,9-10H,3-8,11-16H2;/q+1;-1
SMILES F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](On2nnc3ccccc23)(N4CCCC4)N5CCCC5
IUPAC Name benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
Molecular Weight (g/mol) 520.39
Formula Weight 520.1704
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.