missing translation for 'onlineSavingsMsg'
Learn More

Benzotriazole, TRC

Product Code. 30338286
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338286 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338286 Supplier Toronto Research Chemicals Supplier No. TRCB20695010G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Benzotriazole
CAS 95-14-7
Purity Grade Notes HPLC
Molecular Formula C6 H5 N3
Quantity 10 g
Synonym 1,2,3-1H-Benzotriazole, 1,2,3-Benzotriazole, 1,2,3-Triaza-1H-indene, 1,2,3-Triazaindene, 1,2-Aminoazophenylene, 1H-1,2,3-Benzotriazole, 1H-Benzo[d]-1,2,3-triazole, 2,3-Diazaindole, Azimidobenzene, Aziminobenzene, B 0094, BLS 1326, BT 120, BT 120 (lubricant additive), BT 120SG, BTA, Benzene azimide, Benzisotriazole, Benzotriazole R, C.V.I. Liquid, CVI, Cobratec 35G, Cobratec 99, D 32-108, Entek, Fujilite BT 20NM, HY 2, ISK 3, Irgamet BTZ, Irgastab I 489, Kemitec TT, LT 02, M 318, Miracle HP 16, NSC 3058, Preventol BZT, RC 8239, Rusmin R, Seetec BT, Seetec BT-R, T 706, TH-BTA, Verzone Crystal, Verzone Crystal 120
InChI Formula InChI=1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)
SMILES c1ccc2[nH]nnc2c1
IUPAC Name 1H-benzotriazole
Molecular Weight (g/mol) 119.12
Formula Weight 119.0483
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.