missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl chloride, 98+%, ACS reagent

Product Code. 10157080 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157080 5 g Glass bottle 5g
10117130 500 g Glass bottle 500g
10157030 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10157080 Supplier Thermo Scientific Acros Supplier No. 402030050
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-88-4 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00000653
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -1.0°C
Density 1.2100g/mL
Boiling Point 198.0°C
Flash Point 93°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5COCl
Refractive Index 1.5520 to 1.5540
Quantity 5 g
Beilstein 09, 182
Fieser 01,50; 05,24; 13,219
Merck Index 15, 1115
Specific Gravity 1.21
Solubility Information Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
Viscosity 0.0012 Pas (30°C)
Formula Weight 140.57
Percent Purity 98+%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Benzoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.