missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl chloride, 99%, pure, AcroSeal™

Código de producto. 11984971 Tienda Thermo Scientific Chemicals Productos
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
missing translation for 'unitSize':
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging missing translation for 'unitSize'
11984971 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11984971 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 220271000
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 98-88-4 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 98-88-4
Melting Point -1.0°C
Density 1.2100g/mL
Boiling Point 198.0°C
Flash Point 93°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C7H5ClO
Linear Formula C6H5COCl
Refractive Index 1.5520 to 1.5540
Quantity 100 mL
Beilstein 09,182
Fieser 01,50; 05,24; 13,219
Merck Index 15,1115
Specific Gravity 1.21
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
Solubility Information Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)Cl
IUPAC Name benzoyl chloride
Molecular Weight (g/mol) 140.57
PubChem CID 7412
ChEBI CHEBI:82275
Viscosity 0.0012 Pas (30°C)
Formula Weight 140.57
Percent Purity 99%
Grade Pure
Chemical Name or Material Benzoyl chloride
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Benzoyl chloride
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.